Product Details

SNP ID
rs146331654
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128309687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGCTCTGCAGCTACCTGAGGGG[C/T]AGCAGCCCGGATAGCATCCTGGATG
Phenotype
MIM: 610727
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRUB2 PubMed Links

Gene Details

Gene
TRUB2
Gene Name
TruB pseudouridine synthase family member 2
There are no transcripts associated with this gene.

View Full Product Details