Product Details

SNP ID
rs146334777
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:83627394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTACATCCTGGGAACTGGTAGGTG[C/T]TCAATAAAGAAATGAAATGTAGGTG
Phenotype
MIM: 611343
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
IDNK PubMed Links

Gene Details

Gene
IDNK
Gene Name
IDNK, gluconokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001551.3 2637 Intron NP_001001551.2
NM_001256915.1 2637 Intron NP_001243844.1
XM_006717110.3 2637 Intron XP_006717173.1
XM_006717111.3 2637 UTR 5 XP_006717174.1
XM_017014719.1 2637 Intron XP_016870208.1
XM_017014720.1 2637 Intron XP_016870209.1
XM_017014721.1 2637 Intron XP_016870210.1
XM_017014722.1 2637 Intron XP_016870211.1
XM_017014723.1 2637 Intron XP_016870212.1
XM_017014724.1 2637 UTR 5 XP_016870213.1
XM_017014725.1 2637 Intron XP_016870214.1

View Full Product Details