Product Details

SNP ID
rs146365190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122241891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGTAATTCACAACCATCATCTT[C/T]GAGATTTCCATGCTGTTCTAGAATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM18 PubMed Links

Gene Details

Gene
RBM18
Gene Name
RNA binding motif protein 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033117.3 732 Missense Mutation CAA,CGA Q189R NP_149108.1
XM_006717319.1 732 Missense Mutation CAA,CGA Q189R XP_006717382.1

View Full Product Details