Product Details

SNP ID
rs146528887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136376273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGCGCCAAAGAGCCGAGCAGAG[A/G]CCAAGGCCCCCTGCCATCTGGACAC
Phenotype
MIM: 607212 MIM: 602777
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CARD9 PubMed Links

Gene Details

Gene
CARD9
Gene Name
caspase recruitment domain family member 9
There are no transcripts associated with this gene.

Gene
SNAPC4
Gene Name
small nuclear RNA activating complex polypeptide 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005266096.2 Intron XP_005266153.1
XM_006717241.2 Intron XP_006717304.1
XM_006717242.3 Intron XP_006717305.1
XM_006717244.2 Intron XP_006717307.1
XM_017015058.1 Intron XP_016870547.1

View Full Product Details