Product Details

SNP ID
rs146803572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:130277317 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGCCTGCTCGGAGGGACTGCAC[A/G]CCACATCTGCCCAGCTGTTTGGCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HMCN2 PubMed Links

Gene Details

Gene
HMCN2
Gene Name
hemicentin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291815.1 Intron NP_001278744.1
XM_011518465.2 Intron XP_011516767.1
XM_011518466.2 Intron XP_011516768.1
XM_011518467.2 Intron XP_011516769.1
XM_011518468.2 Intron XP_011516770.1
XM_011518469.2 Intron XP_011516771.1
XM_011518470.2 Intron XP_011516772.1
XM_017014585.1 Intron XP_016870074.1
XM_017014586.1 Intron XP_016870075.1

View Full Product Details