Product Details
- SNP ID
-
rs146820764
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:133636498 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCGCACTGCAGGGCTCGGCTCCC[C/T]GTGAGAGCCCCCTCCCCTATCACAT
- Phenotype
-
MIM: 609312
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DBH
PubMed Links
Gene Details
- Gene
- DBH
- Gene Name
- dopamine beta-hydroxylase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000787.3 |
136 |
Missense Mutation |
CGT,TGT |
R43C |
NP_000778.3 |
View Full Product Details