Product Details

SNP ID
rs146891886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97601739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCAATAAACTATACAGTTGACCC[C/T]TGAACAATATGGGTTTGAACTGCAT
Phenotype
MIM: 190930
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMOD1 PubMed Links

Gene Details

Gene
TMOD1
Gene Name
tropomodulin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166116.1 3334 UTR 3 NP_001159588.1
NM_003275.3 3334 UTR 3 NP_003266.1
Gene
TSTD2
Gene Name
thiosulfate sulfurtransferase like domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139246.4 3334 UTR 3 NP_640339.4
XM_011518312.2 3334 UTR 3 XP_011516614.1
XM_017014342.1 3334 UTR 3 XP_016869831.1

View Full Product Details