Product Details

SNP ID
rs146953824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:26905740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTGACAAACATTGGGCTTTCCCT[C/T]CAATGTTATGGTCTTTATGAAAACA
Phenotype
MIM: 603873
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLAA PubMed Links

Gene Details

Gene
PLAA
Gene Name
phospholipase A2 activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031689.2 2518 Missense Mutation GAA,GGA E720G NP_001026859.1
NM_001321546.1 2518 Missense Mutation GAA,GGA E697G NP_001308475.1
XM_011518072.2 2518 Intron XP_011516374.1
XM_017015319.1 2518 Intron XP_016870808.1

View Full Product Details