Product Details

SNP ID
rs147012546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122856100 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTGTCCTTTAAGTGCTCATATG[A/G]AACATTTGACTAATACTATCAAAAC
Phenotype
MIM: 615231
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RC3H2 PubMed Links

Gene Details

Gene
RC3H2
Gene Name
ring finger and CCCH-type domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100588.1 Intron NP_001094058.1
NM_018835.2 Intron NP_061323.2

View Full Product Details