Product Details

SNP ID
rs147030133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:130904153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCAGGAGGACCACTCCACGCCGT[C/T]GGCATAGGAGGCGTGCGCACCGCGC
Phenotype
MIM: 613357 MIM: 609795
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FIBCD1 PubMed Links

Gene Details

Gene
FIBCD1
Gene Name
fibrinogen C domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145106.1 1404 Missense Mutation AAC,GAC N433D NP_001138578.1
NM_032843.4 1404 Missense Mutation AAC,GAC N433D NP_116232.3
Gene
QRFP
Gene Name
pyroglutamylated RFamide peptide
There are no transcripts associated with this gene.

View Full Product Details