Product Details

SNP ID
rs147168186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35735136 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGAACAAATTCTGAAACGTGTG[C/T]GGAGGAAGATTCGAAATAAAAGATC
Phenotype
MIM: 606443 MIM: 609471 MIM: 186745
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CREB3 PubMed Links

Gene Details

Gene
CREB3
Gene Name
cAMP responsive element binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006368.4 916 Missense Mutation CGG,TGG R155W NP_006359.3
Gene
GBA2
Gene Name
glucosylceramidase beta 2
There are no transcripts associated with this gene.

Gene
MIR6853
Gene Name
microRNA 6853
There are no transcripts associated with this gene.

Gene
TLN1
Gene Name
talin 1
There are no transcripts associated with this gene.

View Full Product Details