Product Details

SNP ID
rs147366726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:841842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCACTTCTCCTAGGGGCACCATG[A/C]CCAACGACGAGGCATTCAGCAAGCC
Phenotype
MIM: 602424
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DMRT1 PubMed Links

Gene Details

Gene
DMRT1
Gene Name
doublesex and mab-3 related transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021951.2 196 Missense Mutation ACC,CCC T2P NP_068770.2
XM_006716732.1 196 Missense Mutation ACC,CCC T2P XP_006716795.1
XM_011517770.1 196 Intron XP_011516072.1
XM_011517771.1 196 Intron XP_011516073.1
XM_011517772.1 196 Intron XP_011516074.1
XM_011517773.2 196 Intron XP_011516075.1
XM_017014374.1 196 Intron XP_016869863.1
XM_017014375.1 196 Missense Mutation ACC,CCC T2P XP_016869864.1

View Full Product Details