Product Details

SNP ID
rs147508822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92327040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCACAAATGAGAGTAAATGTATT[A/G]AAGACCCTTGCTATTTGAGGTGTGC
Phenotype
MIM: 611505 MIM: 611534
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CENPP PubMed Links

Gene Details

Gene
CENPP
Gene Name
centromere protein P
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012267.2 Intron NP_001012267.1
NM_001286969.1 Intron NP_001273898.1
NM_001286971.1 Intron NP_001273900.1
XM_011518685.2 Intron XP_011516987.1
XM_011518689.1 Intron XP_011516991.1
XM_017014715.1 Intron XP_016870204.1
Gene
NOL8
Gene Name
nucleolar protein 8
There are no transcripts associated with this gene.

View Full Product Details