Product Details

SNP ID
rs147544712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120905181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGGACGCTGAGCTTAGGTCAG[G/T]CCGGAAGGCGTCAATGGCGTGCTCA
Phenotype
MIM: 609740 MIM: 601711
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PHF19 PubMed Links

Gene Details

Gene
PHF19
Gene Name
PHD finger protein 19
There are no transcripts associated with this gene.

Gene
TRAF1
Gene Name
TNF receptor associated factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190945.1 1515 Missense Mutation ACT,CCT T364P NP_001177874.1
NM_001190947.1 1515 Missense Mutation ACT,CCT T242P NP_001177876.1
NM_005658.4 1515 Missense Mutation ACT,CCT T364P NP_005649.1

View Full Product Details