Product Details

SNP ID
rs147601195
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:105560160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTATTTATTCAACAAATATTTAT[A/G]TGTTTCAAGCACTGCTGGTTCTAGG
Phenotype
MIM: 607440 MIM: 609829
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FKTN PubMed Links

Gene Details

Gene
FKTN
Gene Name
fukutin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079802.1 Intron NP_001073270.1
NM_001198963.1 Intron NP_001185892.1
NM_006731.2 Intron NP_006722.2
XM_006717014.2 Intron XP_006717077.1
XM_011518368.2 Intron XP_011516670.1
XM_011518369.2 Intron XP_011516671.1
XM_011518370.2 Intron XP_011516672.1
XM_011518371.2 Intron XP_011516673.1
XM_011518373.2 Intron XP_011516675.1
XM_011518374.2 Intron XP_011516676.1
XM_011518375.2 Intron XP_011516677.1
XM_011518376.2 Intron XP_011516678.1
XM_011518378.2 Intron XP_011516680.1
XM_011518379.2 Intron XP_011516681.1
XM_011518381.2 Intron XP_011516683.1
XM_011518387.2 Intron XP_011516689.1
XM_011518389.2 Intron XP_011516691.1
XM_011518390.2 Intron XP_011516692.1
XM_011518391.2 Intron XP_011516693.1
XM_017014462.1 Intron XP_016869951.1
XM_017014463.1 Intron XP_016869952.1
XM_017014464.1 Intron XP_016869953.1
XM_017014465.1 Intron XP_016869954.1
XM_017014466.1 Intron XP_016869955.1
XM_017014467.1 Intron XP_016869956.1
XM_017014468.1 Intron XP_016869957.1
XM_017014469.1 Intron XP_016869958.1
XM_017014470.1 Intron XP_016869959.1
XM_017014471.1 Intron XP_016869960.1
XM_017014472.1 Intron XP_016869961.1
XM_017014473.1 Intron XP_016869962.1
XM_017014474.1 Intron XP_016869963.1
XM_017014475.1 Intron XP_016869964.1
XM_017014476.1 Intron XP_016869965.1
XM_017014477.1 Intron XP_016869966.1
XM_017014478.1 Intron XP_016869967.1
XM_017014479.1 Intron XP_016869968.1
Gene
FSD1L
Gene Name
fibronectin type III and SPRY domain containing 1 like
There are no transcripts associated with this gene.

View Full Product Details