Product Details

SNP ID
rs147653832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37126385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGCATACGAAGATGATCTTTATC[A/G]AGATGAGTCATCTAGTGAACTGAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZCCHC7 PubMed Links

Gene Details

Gene
ZCCHC7
Gene Name
zinc finger CCHC-type containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289119.1 226 Missense Mutation CAA,CGA Q18R NP_001276048.1
NM_001289120.1 226 Missense Mutation CAA,CGA Q18R NP_001276049.1
NM_001289121.1 226 Missense Mutation CAA,CGA Q18R NP_001276050.1
NM_032226.2 226 Missense Mutation CAA,CGA Q18R NP_115602.2
XM_005251608.4 226 Missense Mutation CAA,CGA Q18R XP_005251665.1
XM_005251612.2 226 Missense Mutation CAA,CGA Q18R XP_005251669.1
XM_011518050.1 226 Missense Mutation CAA,CGA Q18R XP_011516352.1
XM_011518051.2 226 Intron XP_011516353.1
XM_017015197.1 226 Missense Mutation CAA,CGA Q18R XP_016870686.1
XM_017015198.1 226 Missense Mutation CAA,CGA Q18R XP_016870687.1
XM_017015199.1 226 UTR 5 XP_016870688.1
XM_017015200.1 226 UTR 5 XP_016870689.1
XM_017015201.1 226 Intron XP_016870690.1

View Full Product Details