Product Details
- SNP ID
-
rs147739827
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:75066790 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGCTAGTTCTTGTATTAGATCTT[C/T]ATATACTTGCAAAAAGAGGTCCTCT
- Phenotype
-
MIM: 608704
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NMRK1
PubMed Links
Gene Details
- Gene
- NMRK1
- Gene Name
- nicotinamide riboside kinase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001127603.1 |
1705 |
Missense Mutation |
AAA,GAA |
K159E |
NP_001121075.1 |
NM_017881.2 |
1705 |
Missense Mutation |
AAA,GAA |
K183E |
NP_060351.1 |
XM_006717162.1 |
1705 |
Missense Mutation |
AAA,GAA |
K187E |
XP_006717225.1 |
XM_006717163.2 |
1705 |
Missense Mutation |
AAA,GAA |
K183E |
XP_006717226.1 |
XM_011518817.1 |
1705 |
Missense Mutation |
AAA,GAA |
K187E |
XP_011517119.1 |
XM_011518818.1 |
1705 |
Missense Mutation |
AAA,GAA |
K187E |
XP_011517120.1 |
XM_011518819.2 |
1705 |
Missense Mutation |
AAA,GAA |
K163E |
XP_011517121.1 |
XM_017014868.1 |
1705 |
Missense Mutation |
AAA,GAA |
K187E |
XP_016870357.1 |
XM_017014869.1 |
1705 |
Missense Mutation |
AAA,GAA |
K187E |
XP_016870358.1 |
XM_017014870.1 |
1705 |
Missense Mutation |
AAA,GAA |
K183E |
XP_016870359.1 |
XM_017014871.1 |
1705 |
Missense Mutation |
AAA,GAA |
K163E |
XP_016870360.1 |
XM_017014872.1 |
1705 |
Missense Mutation |
AAA,GAA |
K163E |
XP_016870361.1 |
XM_017014873.1 |
1705 |
Missense Mutation |
AAA,GAA |
K163E |
XP_016870362.1 |
XM_017014874.1 |
1705 |
Missense Mutation |
AAA,GAA |
K159E |
XP_016870363.1 |
View Full Product Details