Product Details

SNP ID
rs148037657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:71912227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCAGCTACGATTTTTTTTTCCA[A/G]AAACCCCAAGCCCATAGAGGAAATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABHD17B PubMed Links

Gene Details

Gene
ABHD17B
Gene Name
abhydrolase domain containing 17B
There are no transcripts associated with this gene.

Gene
C9orf85
Gene Name
chromosome 9 open reading frame 85
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182505.3 Intron NP_872311.2
XM_011518226.2 Intron XP_011516528.1

View Full Product Details