Product Details

SNP ID
rs148170396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:101362577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTCGTTGAGGCACAGCACAGAG[A/G]AGAATAGGGAGGTTCTATCAGGTGG
Phenotype
MIM: 602938
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAAT PubMed Links

Gene Details

Gene
BAAT
Gene Name
bile acid-CoA:amino acid N-acyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127610.1 1318 Missense Mutation CCT,TCT P370S NP_001121082.1
NM_001701.3 1318 Missense Mutation CCT,TCT P370S NP_001692.1

View Full Product Details