Product Details

SNP ID
rs149119376
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:130447239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTGGAAGGCCTAGTGCAGGGAA[A/G]AGCAAGTGGCCTGCAGTCAGGCTGG
Phenotype
MIM: 603470
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASS1 PubMed Links

Gene Details

Gene
ASS1
Gene Name
argininosuccinate synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000050.4 Intron NP_000041.2
NM_054012.3 Intron NP_446464.1
XM_005272200.3 Intron XP_005272257.1
XM_011518705.2 Intron XP_011517007.1
XM_017014729.1 Intron XP_016870218.1

View Full Product Details