Product Details

SNP ID
rs149172628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137169532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGCGCGCGCAGCGGCGCGAAAG[C/T]CCCTGGTGCCAGTGCTTCCAGCTGG
Phenotype
MIM: 606946 MIM: 138249 MIM: 613505
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANAPC2 PubMed Links

Gene Details

Gene
ANAPC2
Gene Name
anaphase promoting complex subunit 2
There are no transcripts associated with this gene.

Gene
GRIN1
Gene Name
glutamate ionotropic receptor NMDA type subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000832.6 508 Intron NP_000823.4
NM_001185090.1 508 Intron NP_001172019.1
NM_001185091.1 508 Intron NP_001172020.1
NM_007327.3 508 Intron NP_015566.1
NM_021569.3 508 Intron NP_067544.1
XM_005266071.3 508 Intron XP_005266128.1
XM_005266072.3 508 Intron XP_005266129.1
XM_005266073.4 508 Intron XP_005266130.1
XM_011518583.2 508 Intron XP_011516885.1
Gene
LRRC26
Gene Name
leucine rich repeat containing 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013653.2 508 Missense Mutation NP_001013675.1
Gene
MIR3621
Gene Name
microRNA 3621
There are no transcripts associated with this gene.

Gene
TMEM210
Gene Name
transmembrane protein 210
There are no transcripts associated with this gene.

View Full Product Details