Product Details

SNP ID
rs149274311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37917476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATGGTCTACAGGGAAGGACCGT[A/G]AGGTGCGGCCCCACCCAGCAGCCTC
Phenotype
MIM: 600314
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SHB PubMed Links

Gene Details

Gene
SHB
Gene Name
SH2 domain containing adaptor protein B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003028.2 4443 UTR 3 NP_003019.2

View Full Product Details