Product Details

SNP ID
rs149886991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33934351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATACAACTCAGATATGTGCTTGG[A/C]AATAAAACATCATTGTCTTTTCCAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SNORD121B PubMed Links

Gene Details

Gene
SNORD121B
Gene Name
small nucleolar RNA, C/D box 121B
There are no transcripts associated with this gene.

Gene
UBAP2
Gene Name
ubiquitin associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282529.1 14 Intron NP_001269458.1
NM_001282530.1 14 UTR 5 NP_001269459.1
NM_018449.3 14 Intron NP_060919.3

View Full Product Details