Product Details
- SNP ID
-
rs149961183
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:124019593 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGTAGTATTTGAATTTCAGGTAA[A/G]CAACAAATACATTTTTAGTATAGGT
- Phenotype
-
MIM: 603759
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LHX2
PubMed Links
Gene Details
- Gene
- LHX2
- Gene Name
- LIM homeobox 2
View Full Product Details