Product Details
- SNP ID
-
rs150127578
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:32784502 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCAGACCTAGAATCCGGCAAGGGG[A/G]GCTCAGATGAGCTGGCTAAGAAGGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM215
PubMed Links
Gene Details
- Gene
- TMEM215
- Gene Name
- transmembrane protein 215
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_212558.2 |
684 |
Missense Mutation |
AGC,GGC |
S107G |
NP_997723.2 |
View Full Product Details