Product Details

SNP ID
rs150505158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:21239874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTGAGACAGATTACAGCCCAGA[G/T]AGCAGCTTGACTTGCAGCTGAGCAC
Phenotype
MIM: 147579
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IFNA14 PubMed Links

Gene Details

Gene
IFNA14
Gene Name
interferon alpha 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002172.2 106 Missense Mutation TAT,TCT Y21S NP_002163.2

View Full Product Details