Product Details

SNP ID
rs150559384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:131009573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCAGTACCCCACCTCGGTCAACA[C/T]CACCCTCCGCCTAGGTAAGCGCGGG
Phenotype
MIM: 604349
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAMC3 PubMed Links

Gene Details

Gene
LAMC3
Gene Name
laminin subunit gamma 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006059.3 457 Missense Mutation ACC,ATC T120I NP_006050.3
XM_006716921.2 457 Missense Mutation ACC,ATC T120I XP_006716984.1
XM_011518121.1 457 Missense Mutation ACC,ATC T120I XP_011516423.1

View Full Product Details