Product Details

SNP ID
rs150578575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35042358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGCAGACACTGCGGCAGCAGCT[A/G]CTTCCAGAGTCCAGGAAATCTGGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf131 PubMed Links

Gene Details

Gene
C9orf131
Gene Name
chromosome 9 open reading frame 131
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040410.2 151 Missense Mutation TAC,TGC Y35C NP_001035500.1
NM_001040411.2 151 Intron NP_001035501.1
NM_001040412.2 151 Intron NP_001035502.1
NM_001287391.1 151 Intron NP_001274320.1
NM_203299.3 151 Missense Mutation TAC,TGC Y35C NP_976044.2

View Full Product Details