Product Details

SNP ID
rs150763272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:101540286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGCGGCAGGCCATTGAAGATGA[A/G]CTTCGTGAGCACATTGAAAAACTGG
Phenotype
MIM: 607699
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF20 PubMed Links

Gene Details

Gene
RNF20
Gene Name
ring finger protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019592.6 582 Silent Mutation GAA,GAG E71E NP_062538.5
XM_011518862.1 582 Silent Mutation GAA,GAG E71E XP_011517164.1

View Full Product Details