Product Details

SNP ID
rs150962629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:105662800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAACTACCAGGTTCCTTCACCT[C/G]GTCCAAGCCACCACATTCCTTAGTG
Phenotype
MIM: 607440 MIM: 186855
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FKTN PubMed Links

Gene Details

Gene
FKTN
Gene Name
fukutin
There are no transcripts associated with this gene.

Gene
TAL2
Gene Name
TAL bHLH transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005421.2 344 Missense Mutation CGT,GGT R102G NP_005412.1

View Full Product Details