Product Details

SNP ID
rs151241381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35660646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCAGCTGGACCCCCTTTTTGAG[C/T]GGTGAGGAGAGCAATGATTCTGTGA
Phenotype
MIM: 157660 MIM: 604964
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF39 PubMed Links

Gene Details

Gene
ARHGEF39
Gene Name
Rho guanine nucleotide exchange factor 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032818.2 2108 UTR 3 NP_116207.2
XM_011518057.2 2108 UTR 3 XP_011516359.1
XM_017015225.1 2108 UTR 3 XP_016870714.1
XM_017015226.1 2108 Intron XP_016870715.1
XM_017015227.1 2108 UTR 3 XP_016870716.1
XM_017015228.1 2108 Intron XP_016870717.1
Gene
CCDC107
Gene Name
coiled-coil domain containing 107
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195200.1 2108 Missense Mutation CGT,TGT R137C NP_001182129.1
NM_001195201.1 2108 Missense Mutation CGC,TGC R137C NP_001182130.1
NM_001195217.1 2108 Silent Mutation CGG,TGG R137W NP_001182146.1
NM_174923.2 2108 Missense Mutation CGT,TGT R137C NP_777583.2
XM_005251403.4 2108 Missense Mutation CGC,TGC R137C XP_005251460.1
Gene
RMRP
Gene Name
RNA component of mitochondrial RNA processing endoribonuclease
There are no transcripts associated with this gene.

Gene
SIT1
Gene Name
signaling threshold regulating transmembrane adaptor 1
There are no transcripts associated with this gene.

View Full Product Details