Product Details

SNP ID
rs112016403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:21656406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCCCCGCGTGACCGTGCAGAGC[C/T]CGCGGTAGTGCCCCAGCCGCCGTCC
Phenotype
MIM: 300724
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNKSR2 PubMed Links

Gene Details

Gene
CNKSR2
Gene Name
connector enhancer of kinase suppressor of Ras 2
There are no transcripts associated with this gene.

Gene
KLHL34
Gene Name
kelch like family member 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153270.2 1982 Silent Mutation CGA,CGG R461R NP_695002.1

View Full Product Details