Product Details

SNP ID
rs112616756
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153949138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGCTCCTCTCTCTGCTTTCCCAT[C/G]TGCCTCTGCTTCCTCCCAACAGCAA
Phenotype
MIM: 300019 MIM: 312420
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HCFC1 PubMed Links

Gene Details

Gene
HCFC1
Gene Name
host cell factor C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005334.2 7319 UTR 3 NP_005325.2
XM_006724815.2 7319 UTR 3 XP_006724878.1
XM_006724816.2 7319 UTR 3 XP_006724879.1
XM_011531147.2 7319 UTR 3 XP_011529449.1
XM_011531148.2 7319 UTR 3 XP_011529450.1
XM_017029471.1 7319 UTR 3 XP_016884960.1
XM_017029472.1 7319 UTR 3 XP_016884961.1
Gene
RENBP
Gene Name
renin binding protein
There are no transcripts associated with this gene.

View Full Product Details