Product Details

SNP ID
rs138204346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71301843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCGTAACAAAGGCTGTGGGCAG[C/T]ACTTTGACCCTAATACCAACCTTCC
Phenotype
MIM: 300332 MIM: 300084
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITGB1BP2 PubMed Links

Gene Details

Gene
ITGB1BP2
Gene Name
integrin subunit beta 1 binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303277.1 96 UTR 5 NP_001290206.1
NM_012278.2 96 Missense Mutation CAC,TAC H13Y NP_036410.1
XM_017029405.1 96 UTR 5 XP_016884894.1
Gene
NONO
Gene Name
non-POU domain containing, octamer-binding
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145408.1 96 Intron NP_001138880.1
NM_001145409.1 96 Intron NP_001138881.1
NM_001145410.1 96 Intron NP_001138882.1
NM_007363.4 96 Intron NP_031389.3

View Full Product Details