Product Details

SNP ID
rs138209054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130384483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGAGATAGTGACGATGTGACAG[C/G]AACTTTCCCTGGGCCTCTCTGGGCC
Phenotype
MIM: 300513
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPR119 PubMed Links

Gene Details

Gene
GPR119
Gene Name
G protein-coupled receptor 119
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178471.2 1055 Missense Mutation TCC,TGC S322C NP_848566.1

View Full Product Details