Product Details

SNP ID
rs138227292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154485870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGACCTGCGGGGGTGGGGAGTC[G/T]GCCAGCCTCTGGGCCTCGCCTTCTT
Phenotype
MIM: 300060 MIM: 312090 MIM: 312070
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LAGE3 PubMed Links

Gene Details

Gene
LAGE3
Gene Name
L antigen family member 3
There are no transcripts associated with this gene.

Gene
SLC10A3
Gene Name
solute carrier family 10 member 3
There are no transcripts associated with this gene.

Gene
UBL4A
Gene Name
ubiquitin like 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014235.4 350 Silent Mutation GCA,GCC A88A NP_055050.1

View Full Product Details