Product Details

SNP ID
rs138298502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135347457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAGACTTACAGAGAGAACTTGA[A/G]ATACCAGAGCAGCAGGTAAGAAAAG
Phenotype
MIM: 300627 MIM: 314997
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF449 PubMed Links

Gene Details

Gene
ZNF449
Gene Name
zinc finger protein 449
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152695.5 504 Silent Mutation GAA,GAG E113E NP_689908.3
XM_011531312.2 504 Silent Mutation GAA,GAG E113E XP_011529614.1
XM_011531313.2 504 Intron XP_011529615.1
XM_011531314.2 504 UTR 5 XP_011529616.1
XM_017029350.1 504 UTR 5 XP_016884839.1
XM_017029351.1 504 Intron XP_016884840.1
XM_017029352.1 504 Intron XP_016884841.1
Gene
ZNF75D
Gene Name
zinc finger protein 75D
There are no transcripts associated with this gene.

View Full Product Details