Product Details

SNP ID
rs138329093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:115005268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATAACTAATATTAAGATGAAAC[C/T]AAATGGTAGCCAGAAACGTAGCAAA
Phenotype
MIM: 300130
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL13RA2 PubMed Links

Gene Details

Gene
IL13RA2
Gene Name
interleukin 13 receptor subunit alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000640.2 1170 Missense Mutation NP_000631.1

View Full Product Details