Product Details

SNP ID
rs138359911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:151717364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGGCTCTGCTTCAGCCAAACGA[C/G]TTTGGAATATGACTGCCACCCGACC
Phenotype
MIM: 300450
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FATE1 PubMed Links

Gene Details

Gene
FATE1
Gene Name
fetal and adult testis expressed 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033085.2 283 Missense Mutation CTT,GTT L67V NP_149076.1

View Full Product Details