Product Details

SNP ID
rs138364468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:13574806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAAAAGTGAAAAGTACATGCTG[A/G]AGTACAAGAAGACTCTGAAGATGAT
Phenotype
MIM: 300239
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EGFL6 PubMed Links

Gene Details

Gene
EGFL6
Gene Name
EGF like domain multiple 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167890.1 Intron NP_001161362.1
NM_015507.3 Intron NP_056322.2

View Full Product Details