Product Details

SNP ID
rs138411334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:12799294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTAACGACAACCTGATGGAACT[C/T]CTCATCATGATCAATGCCTGCAAGA
Phenotype
MIM: 311860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRPS2 PubMed Links

Gene Details

Gene
PRPS2
Gene Name
phosphoribosyl pyrophosphate synthetase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039091.2 353 Silent Mutation CTC,CTT L70L NP_001034180.1
NM_002765.4 353 Silent Mutation CTC,CTT L70L NP_002756.1

View Full Product Details