Product Details

SNP ID
rs138422269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:118741142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATGGTATTTTAGTCATTTTGGC[A/G]ACAAACAAGATAAGGTAAGTTTTCT
Phenotype
MIM: 300119
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL13RA1 PubMed Links

Gene Details

Gene
IL13RA1
Gene Name
interleukin 13 receptor subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001560.2 413 Missense Mutation AAC,GAC N72D NP_001551.1
XM_011531336.1 413 Missense Mutation AAC,GAC N174D XP_011529638.1
XM_011531337.1 413 Intron XP_011529639.1
XM_017029507.1 413 UTR 5 XP_016884996.1

View Full Product Details