Product Details

SNP ID
rs138513845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48706306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGAGCGGCGGGGCCAGATCTA[C/T]GACCGTCAGGGCGCCACCTACCTCT
Phenotype
MIM: 300254
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUV39H1 PubMed Links

Gene Details

Gene
SUV39H1
Gene Name
suppressor of variegation 3-9 homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282166.1 1169 Silent Mutation TAC,TAT Y301Y NP_001269095.1
NM_003173.3 1169 Silent Mutation TAC,TAT Y290Y NP_003164.1

View Full Product Details