Product Details
- SNP ID
-
rs138687868
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:111246723 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGTCAGCATCCAGAGTAACCTGCC[C/T]CAAGAACTGATCACAGAATTTTCGG
- Phenotype
-
MIM: 300146
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CAPN6
PubMed Links
Gene Details
- Gene
- CAPN6
- Gene Name
- calpain 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014289.3 |
1971 |
Missense Mutation |
AGG,GGG |
R594G |
NP_055104.2 |
View Full Product Details