Product Details

SNP ID
rs138706948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:116172511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTTGTCAAAAGGGTCCTAAAAA[G/T]GTTTCTAGCATATACATCTTCAACC
Phenotype
MIM: 300034
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AGTR2 PubMed Links

Gene Details

Gene
AGTR2
Gene Name
angiotensin II receptor type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000686.4 336 Missense Mutation AAG,AAT K77N NP_000677.2
XM_011537533.1 336 Missense Mutation AAG,AAT K77N XP_011535835.1

View Full Product Details