Product Details

SNP ID
rs138722887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:155212285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCCCAAAGGAGATTCAGGAACA[G/T]GAAAGCAGAGTTTGAACCTTTTTAC
Phenotype
MIM: 300133
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
VBP1 PubMed Links

Gene Details

Gene
VBP1
Gene Name
VHL binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303543.1 Intron NP_001290472.1
NM_001303544.1 Intron NP_001290473.1
NM_001303545.1 Intron NP_001290474.1
NM_003372.6 Intron NP_003363.1

View Full Product Details