Product Details

SNP ID
rs138854192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48479065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCACTGCCAAGGAGATCATCCAG[C/T]ACTTTAAGGGCTGCCCTGCGGACCT
Phenotype
MIM: 300499 MIM: 300649
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FTSJ1 PubMed Links

Gene Details

Gene
FTSJ1
Gene Name
FtsJ RNA methyltransferase homolog 1 (E. coli)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282157.1 728 UTR 5 NP_001269086.1
NM_012280.3 728 Missense Mutation CAC,TAC H104Y NP_036412.1
NM_177439.2 728 Missense Mutation CAC,TAC H104Y NP_803188.1
XM_005272595.1 728 Missense Mutation CAC,TAC H104Y XP_005272652.1
XM_011543893.2 728 Missense Mutation CAC,TAC H104Y XP_011542195.1
XM_011543894.2 728 Missense Mutation CAC,TAC H104Y XP_011542196.1
XM_011543895.1 728 Missense Mutation CAC,TAC H104Y XP_011542197.1
XM_017029382.1 728 Missense Mutation CAC,TAC H104Y XP_016884871.1
Gene
SLC38A5
Gene Name
solute carrier family 38 member 5
There are no transcripts associated with this gene.

View Full Product Details