Product Details

SNP ID
rs139042252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:93672499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTGCAGCCCTTGCCTTAGCCTCT[C/G]TTGCTTTAGGCTGTTCTTTATCATC
Phenotype
MIM: 300117
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM133A PubMed Links

Gene Details

Gene
FAM133A
Gene Name
family with sequence similarity 133 member A
There are no transcripts associated with this gene.

Gene
NAP1L3
Gene Name
nucleosome assembly protein 1 like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004538.5 1185 Missense Mutation ACA,AGA T269R NP_004529.2

View Full Product Details