Product Details

SNP ID
rs139389280
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153767065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACCGCCTCTTCCAGCTCAGCCCC[A/G]AGCTGCAGCTCGAGGCCGTGGCTGT
Phenotype
MIM: 300214
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLXNB3 PubMed Links

Gene Details

Gene
PLXNB3
Gene Name
plexin B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163257.1 412 Missense Mutation AAG,GAG K103E NP_001156729.1
NM_005393.2 412 Missense Mutation AAG,GAG K80E NP_005384.2

View Full Product Details