Product Details

SNP ID
rs139405979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:74304567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCTGCGGCAGACTAGGACATCTG[A/G]CTCGTGACTGTGATCGTCAGAAAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZCCHC13 PubMed Links

Gene Details

Gene
ZCCHC13
Gene Name
zinc finger CCHC-type containing 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203303.2 378 Missense Mutation ACT,GCT T101A NP_976048.1

View Full Product Details